Learn More
Hanna Instruments™ Edge™ pH Electrode
Click to view available options
pH Range:
0 to 12
Unit Size:
Each
Description
HI10530 digital, glass refillable, conical point, Hanna Instruments
Specifications
Specifications
| Connector Types | 3.5 mm |
| Type | pH Electrode |
| For Use With (Application) | Semi-solids and emulsions |
| Internal Reference | Double, Ag/AgCl |
| Junction | Ceramic, Triple |
| Material | Glass Body |
| pH Range | 0 to 12 |
| Temperature (Metric) Operating | -5°C to 70°C |