Learn More
Hanna Instruments™ Edge™ pH Electrode
Description
HI12301 digital, plastic refillable, Hanna Instruments
Specifications
Specifications
| Connector Types | 3.5 mm |
| Filling Solution Type | Gel |
| Internal Reference | Double, Ag/AgCl |
| Junction | Ceramic, Single |
| Material | PEI Body |
| pH Range | 0 to 12 |
| Length (Metric) Cable | 1 m |
| Diameter (Metric) | 7.5 mm |