Learn More
Edonerpic maleate, MedChemExpress
Product Code. 16477406
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16477406
Brand: Medchem Express HY17631A
MedChemExpress Edonerpic maleate is a novel neurotrophic agent which can inhibit amyloid-β peptides (Aβ).
Edonerpic maleate is a novel neurotrophic agent which can inhibit amyloid-β peptides (Aβ).
Specifications
Chemical Name or Material Edonerpic maleate
CAS 519187-97-4
Color White
Purity Grade Notes Research
Molecular Formula C20H25NO6S
Quantity 100 mg
Synonym T-817 maleate T-817MA
Solubility Information DMSO : 100 mg/mL (245.41 mM; Need ultrasonic) ∣H2O : 100 mg/mL (245.41 mM; Need ultrasonic)
SMILES OC1CN(CCCOCCC2=CC=C3C(C=CS3)=C2)C1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 407.48
Formula Weight 407.48
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less
Research purposes only