missing translation for 'onlineSavingsMsg'
Learn More

Edonerpic maleate, MedChemExpress

Product Code. 16409917
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409917

Brand: Medchem Express HY17631A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Edonerpic maleate is a novel neurotrophic agent which can inhibit amyloid-β peptides (Aβ).

Edonerpic maleate is a novel neurotrophic agent which can inhibit amyloid-β peptides (Aβ).

Specifications

Chemical Name or Material Edonerpic maleate
CAS 519187-97-4
Color White
Purity Grade Notes Research
Molecular Formula C20H25NO6S
Quantity 5 mg
Synonym T-817 maleate T-817MA
Solubility Information DMSO : 100 mg/mL (245.41 mM; Need ultrasonic) ∣H2O : 100 mg/mL (245.41 mM; Need ultrasonic)
SMILES OC1CN(CCCOCCC2=CC=C3C(C=CS3)=C2)C1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 407.48
Formula Weight 407.48
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.