missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDRF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568488
This item is not returnable.
View return policy
Description
EDRF1 Polyclonal Antibody for Western Blot
This gene may play a role in erythroid cell differentiation. The encoded protein inhibits DNA binding of the erythroid transcription factor GATA-1 and may regulate the expression of alpha-globin and gamma-globin. Alternative splicing results in multiple transcript variants.
Specifications
| EDRF1 | |
| Polyclonal | |
| Unconjugated | |
| Edrf1 | |
| 2700050L05Rik; AU022667; AW558805; C10orf137; EDRF1; erythroid differentiation regulatory factor 1; erythroid differentiation-related factor 1; RGD1306820 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26098 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5-1.0 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3B7T1 | |
| Edrf1 | |
| synthetic peptide directed towards the Middle region of human EDRF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction