missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEA1 Antibody (6D4.1D4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36568C
This item is not returnable.
View return policy
Description
EEA1 Monoclonal specifically detects EEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| EEA1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 6D4.1D4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| Q15075 | |
| EEA1 | |
| Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075] | |
| 0.1 mL | |
| Endosome Markers, Membrane Trafficking and Chaperones | |
| 8411 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction