missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EED Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98765C
This item is not returnable.
View return policy
Description
EED Polyclonal antibody specifically detects EED in Human samples. It is validated for Western Blot
Specifications
| EED | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| embryonic ectoderm development, HEED, polycomb protein EED, WAIT1, WAIT-1, WD protein associating with integrin cytoplasmic tails 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human EED | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 8726 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction