missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eEF-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16274
This item is not returnable.
View return policy
Description
eEF-2 Polyclonal specifically detects eEF-2 in Human samples. It is validated for Western Blot.
Specifications
| eEF-2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EF-2, EF2EEF-2, elongation factor 2, eukaryotic translation elongation factor 2, polypeptidyl-tRNA translocase | |
| Rabbit | |
| 95 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 1938 | |
| Human | |
| IgG |
| Western Blot | |
| 0.94mg/mL | |
| Western Blot 1:500-1:3000 | |
| P13639 | |
| EEF2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human eEF2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (99%), Chicken (98%), Rat (99%), Bovine (99%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction