missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | EEF1A2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:400 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635112
|
Novus Biologicals
NBP2-97554-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691332
|
Novus Biologicals
NBP2-97554-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EEF1A2 Polyclonal antibody specifically detects EEF1A2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| EEF1A2 | |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Signaling, Oncogenes, Transcription Factors and Regulators | |
| PBS (pH 7.0) | |
| 1917 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| eEF1A-2, EEF1ALFLJ41696, EF1A, EF-1-alpha-2, elongation factor 1-alpha 2, elongation factor-1 alpha, Eukaryotic elongation factor 1 A-2, eukaryotic translation elongation factor 1 alpha 2, HS1, statin, statin S1, statin-like, Statin-S1, STN, STNL | |
| Produced in rabbits immunized with E. coli-derived Human EEF1A2 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel