missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eEF1A2 binding protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48962
This item is not returnable.
View return policy
Description
eEF1A2 binding protein Polyclonal antibody specifically detects eEF1A2 binding protein in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| eEF1A2 binding protein | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| DKFZp434B1231, eEF1A2 binding protein, EEF1A2-binding protein 1, EEF1A2BP1, immunoglobulin-like and fibronectin type III domain containing 1, immunoglobulin-like and fibronectin type III domain-containing protein 1, KY-interacting protein 1, KYIP1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AHSFRIRVAACPQAPGPIHLQENVPGTVTAEWEPSPDEAQDVPLHYAVFTRSSAHGPWHEAADRIYTNRFTLLGILPGHEYHFRVVA | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 91156 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction