Learn More
Invitrogen™ EEF1A2 Polyclonal Antibody

Description
Immunogen sequence: NPATVPFVPI SGWHGDNMLE PSPNMPWFKG WKVERKEGNA SGVSLLEALD TILPPTRPTD KPLRLPLQDV YKIGGIGTVP VGRVETGILR PGMVVTFAPV NITTEVKSVE MHHEALSEAL PGDNVGFNVK NVSVKDIRRG NVCGDSKSDP PQEAAQFTSQ VIILNHPGQI SAGYSPVIDC HTAHIACKFA ELKEKIDRRS GKKLEDNPKS LKSGDAAIVE MVPGKPMCVE SFSQYPPLGR FAVRDMRQTV AVGVIKNVEK KSGGAGKVTK SAQKAQKAGK; Positive Samples: BT-474, MCF7, 293T, NCI-H460, Mouse brain, Mouse heart, Mouse skeletal muscle, Rat heart, Rat skeletal muscle; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | EEF1A2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EEF1A2 |
| Gene Accession No. | P62631, P62632, Q05639 |
| Gene Alias | Eef1a; EEF1A2; eEF1A-2; EEF1AL; EF1A; EF-1-alpha-2; EIEE33; elongation factor 1 A-2; elongation factor 1-alpha 2; Eukaryotic elongation factor 1 A-2; eukaryotic translation elongation factor 1 alpha 2; HS1; MRD38; Ps10; RATPS10; S1; statin S1; Statin-like protein; statin-S1; STN; Stnl; wasted; wst |
| Gene Symbols | EEF1A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.