missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | EEF1B2 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16866253
|
Proteintech
10095-2-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16856253
|
Proteintech
10095-2-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human EEF1B2.
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5′ UTR.Specifications
| EEF1B2 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P24534 | |
| 1933, 363241, 55949 | |
| EEF1B2 Fusion Protein Ag0135 | |
| Primary | |
| -20°C | |
| EEF1B2 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| EEF1B, EEF1B1, EEF1B2, EF 1 beta, EF1B, Elongation factor 1 beta | |
| Eef1b2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title