missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1D Monoclonal Antibody (OTI1B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811675S
This item is not returnable.
View return policy
Description
EEF1D Monoclonal antibody specifically detects EEF1D in Human samples. It is validated for Western BlotSpecifications
| EEF1D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z3I7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 137209 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1B9 | |
| Unconjugated | |
| ZNF572 | |
| antigen NY-CO-4; EEF 1D; Eef1d; EF 1 delta; EF 1D; EF1D; EF-1D; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); FP1047; guanine nucleotide exchange protein; translation elongation factor 1-delta subunit | |
| Full length human recombinant protein of human ZNF572 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction