missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1D Mouse anti-Human, Clone: OTI6B3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811677
This item is not returnable.
View return policy
Description
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.Specifications
| EEF1D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P29692 | |
| EEF1D | |
| Full length human recombit protein of human EEF1D produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6B3 | |
| Unconjugated | |
| EEF1D | |
| EF-1D, EF1D, FP1047 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1936 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction