missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEF1D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102619
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EEF1D.
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
Specifications
| EEF1D | |
| Polyclonal | |
| Unconjugated | |
| EEF1D | |
| antigen NY-CO-4; EEF 1D; Eef1d; EF 1 delta; EF 1D; EF1D; EF-1D; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); FP1047; guanine nucleotide exchange protein; translation elongation factor 1-delta subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1936, 300033, 66656 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29692, P57776, Q68FR9 | |
| EEF1D | |
| A synthesized peptide derived from human EEF1D(Accession P29692), corresponding to amino acid residues P32-V82. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido