missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EEF1E1 Polyclonal Antibody

Product Code. 16517821
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16517821 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16517821 Supplier Invitrogen™ Supplier No. PA590407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAAELSLL EKSLGLSKGN KYSAQGERQI PVLQTNNGPS LTGLTTIAAH LVKQANKEYL LGSTAEEKAI VQQWLEYRVT QVDGHSSKND IHTLLKDLNS; Positive Samples: 293T, HeLa, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a multifunctional protein that localizes both in the cytoplasm and in the nucleus. In the cytoplasm, the encoded protein is an auxiliary component of the macromolecular aminoacyl-tRNA synthase complex. However, its mouse homolog has been shown to translocate to the nucleus in response to DNA damage and plays a positive role in ATM/ATR-mediated p53 activation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1E1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EEF1E1
Gene Accession No. O43324, Q9D1M4
Gene Alias 1110003A02Rik; AIMP3; Aminoacyl tRNA synthetase complex-interacting multifunctional protein 3; ARS-interacting multifunctional protein 3; Eef1e1; Elongation factor p18; eukaryotic translation elongation factor 1 epsilon 1; eukaryotic translation elongation factor 1 epsilon-1; Multisynthase complex auxiliary component p18; multisynthetase complex auxiliary component p18; P18; p18 component of aminoacyl-tRNA synthetase complex
Gene Symbols EEF1E1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human EEF1E1 (NP_0042711).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291057, 66143, 9521
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.