Learn More
Invitrogen™ EEF2 Recombinant Rabbit Monoclonal Antibody (4E2Z5)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542937
Product Code. 17838392
  • € 500.00 / 100µL
Description
Positive test controls include: HeLa, NIH/3T3, Jurkat, Mouse liver, Rat spleen. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: IYGVLNRKRG HVFEESQVAG TPMFVVKAYL PVNESFGFTA DLRSNTGGQA FPQCVFDHWQ ILPGDPFDNS SRPSQVVAET RKRKGLKEGI PALDNFLDKL.
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EEF2
Recombinant Monoclonal
0.33 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P05197, P13639, P58252
EEF2
A synthetic peptide corresponding to a sequence within amino acids 759-858 of human eEF2 (P13639).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
4E2Z5
Unconjugated
EEF2
EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2
Rabbit
Affinity Chromatography
RUO
13629, 1938, 29565
-20°C, Avoid Freeze/Thaw Cycles