missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF2K, p Ser398 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23310
This item is not returnable.
View return policy
Description
EEF2K Polyclonal antibody specifically detects EEF2K in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| EEF2K | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase, calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase, EC 2.7.11, EC 2.7.11.20, eEF-2 kinase, eEF-2Kcalmodulin-dependent protein kinase III, elongation factor-2 kinase, eukaroytic elongation factor 2 kinase, eukaryotic elongation factor 2 kinase, eukaryotic elongation factor-2 kinase, HSU93850, MGC45041 | |
| Phospho-eEF2K (Ser-398) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding Ser-398 in human eEF2K. This sequence is well conserved in rat and mouse eEF2K, and has low homology to other proteins. [UniProt# O00418] | |
| 0.1 mL | |
| Cancer, Hypoxia Signaling, Phospho Specific | |
| 29904 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction