Learn More
Invitrogen™ EEF2K Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5117924
Product Code. 17471583
  • € 476.00 / 100µL
Description
EEF2K Polyclonal Antibody for Western Blot, IP
EEF2K is a highly conserved Serine/Threoning kinase in the calmodulin signaling pathway whose activity is increased in cancers including breast cancer. EEF2K (eukaryotic elongation factor 2 kinase) is a threonine kinase that regulates protein synthesis by controlling the rate of peptide chain elongation. Up activation, by a variety of upstream kinases including AMPK or TRPM7, EEF2K phosphorylates the elongation factor EEF2 at a single site. Then, EEF2K renders EEF2 unable to bind ribosomes and inactive - in turn, the rate of protein synthesis is reduced.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EEF2K
Polyclonal
PBS with 0.03% ProClin 300; pH 7
O00418
EEF2K
E. coli-derived Human EEF2K fragment.
100 μL
Primary
Human
Antibody
IgG
Immunoprecipitation, Western Blot
Unconjugated
EEF2K
C86191; Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase; calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase; calmodulin-dependent protein kinase III; EC 2.7.11.20; eEF-2 kinase; Eef2k; eEF-2K; EF2K; elongation factor 2 kinase; elongation factor-2 kinase; euk; eukaroytic elongation factor 2 kinase; eukaryotic elongation factor 2 kinase; eukaryotic elongation factor-2 kinase; HSU93850; kinase eEF2K; SMEF2K
Rabbit
Antigen affinity chromatography, Protein A
RUO
29904
-20°C, Avoid Freeze/Thaw Cycles