missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEN Antibody (OTI2F5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71279AF647
This item is not returnable.
View return policy
Description
EEN Monoclonal specifically detects EEN in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| EEN | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SH3GL1 | |
| Full length recombinant protein of human EEN (NP_003016) produced in HEK293T cells. | |
| 0.1 mL | |
| 6455.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CNSA1endophilin-2, EEN fusion partner of MLL, EENSH3 domain-containing GRB2-like protein 1, Endophilin-2, extra 11-19 leukemia fusion, Extra eleven-nineteen leukemia fusion gene protein, MGC111371, SH3 domain protein 2B, SH3-containing Grb-2-like 1 protein, SH3D2Bendophilin-A2, SH3-domain GRB2-like 1, SH3P8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction