missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEPD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565812
This item is not returnable.
View return policy
Description
Immunogen sequence: SVEDLVRMDGI NAAFLDRIRH QVFAERSRPP STHTNGGLTF TAKPHPSPTS LSLQSEDLDL PPGGPTQIIS TRPSVEAFGG TRDGRPVLRL ATWNLQGCSV Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
EEPD1 gene ontology annotations related to this gene include DNA repair.
Specifications
| EEPD1 | |
| Polyclonal | |
| Unconjugated | |
| EEPD1 | |
| 2310005P05Rik; AI481005; EEPD1; endonuclease/exonuclease/phosphatase family domain containing 1; endonuclease/exonuclease/phosphatase family domain-containing protein 1; HSPC107; hypothetical protein LOC511767; KIAA1706; KIAA1706 protein; RGD1307401 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L9B9 | |
| EEPD1 | |
| Recombinant Human EEPD1. Recombinant protein control fragment (Product #RP-104973). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction