missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Efalizumab Recombinant Human Monoclonal Antibody (hu1124 (Efalizumab))

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547999
This item is not returnable.
View return policy
Description
Specificity: Binds to human LFA-1.
Humanized IgG1 kappa isotype monoclonal antibody that binds to human CD11a. Efalizumab has a molecular weight of approximately 150 kilodaltons and is produced in a Chinese hamster ovary mammalian cell expression system in a nutrient medium containing the antibiotic gentamicin.
Specifications
| Efalizumab | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Inhibition Assays, Immunohistochemistry (Paraffin) | |
| hu1124 (Efalizumab) | |
| Unconjugated | |
| hu1124 | |
| Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction