missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB4B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92725-0.02ml
This item is not returnable.
View return policy
Description
EFCAB4B Polyclonal antibody specifically detects EFCAB4B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EFCAB4B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Calcium release-activated calcium channel regulator 2A, CRAC channel regulator 2A, CRACR2A, DKFZp686G13246, EF-hand calcium binding domain 4B, EF-hand calcium-binding domain-containing protein 4B, FLJ33046, FLJ33805, MGC4266 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CRACR2A (NP_001138430.1). MAAPDGRVVSRPQRLGQGSGQGPKGSGACLHPLDSLEQKETQEQTSGQLVMLRKAQEFFQTCDAEGKGFIARKDMQRLHKELPLSLEELEDVFDALDADGNGYLTPQEFTTGFSHFFFSQNNPSQEDAGEQVAQRHEEKVYLSRGDEDLGDMGEDEEAQFRMLMDRLGAQKVLEDESDVKQLWLQLKKEEPHLLSNFEDF | |
| 0.02 mL | |
| Neuroscience, Signal Transduction | |
| 84766 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction