missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EFHB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144758
This item is not returnable.
View return policy
Description
Immunogen sequence: KVIPVGYRVA TCLTEKLPRL ITPPEAKKYF NFRYPPAGVE RVFYGRANDP QIAPYLTHGI RSKISILANT LINPQPITTF QQKIKDKKES IYLSNRRAPL Antigen sequence identity: Mouse 89%; Rat 81%.
EFHB contains two EF-hand domains. There are three named isoforms.
Specifications
| EFHB | |
| Polyclonal | |
| Unconjugated | |
| EFHB | |
| CFAP21; cilia and flagella associated protein 21; EF-hand domain family member B; EF-hand domain family, member B; EF-hand domain-containing family member B; EFHB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 151651 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7U6 | |
| EFHB | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 266-365. Recombinant protein control fragment (Product #RP-108699). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction