missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EFR3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107118
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 92% Immunogen sequence: PQLTDEDRLS KRRSIGETIS LQVEVESRNS PEKEERVPAE EITYETLKKA.
Component of a complex required to localize phosphatidylinositol 4-kinase (PI4K) to the plasma membrane. The complex acts as a regulator of phosphatidylinositol 4-phosphate (PtdIns(4)P) synthesis (Probable). In the complex, EFR3B probably acts as the membrane-anchoring component. Also involved in responsiveness to G-protein-coupled receptors; it is however unclear whether this role is direct or indirect.
Specifications
| EFR3B | |
| Polyclonal | |
| Unconjugated | |
| EFR3B | |
| AI852640; C030014M07Rik; EFR3 homolog B; EFR3 homolog B (S. cerevisiae); Efr3b; FLJ37871; Kiaa0953; mKIAA0953; Protein EFR3 homolog B; RGD1310845 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22979 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9Y2G0 | |
| EFR3B | |
| Synthetic peptide directed towards the C-terminal region of Human EFR3B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction