missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EG 00229 trifluoroacetate
GREENER_CHOICE

Product Code. 18719296
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18719296

Brand: Tocris Bioscience™ 6986/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neuropilin 1 (NRP1) receptor antagonist; inhibits VEGFA binding to NRP1

EG 00229 trifluoroacetate is a neuropilin 1 (NRP1) antagonist; inhibits VEGFA binding to the NRP1 b1 CendR binding pocket. Has no effect on VEGFA binding to VEGFR-1 and VEGFR-2. Reduces VEGFA-induced VEGFR-2 tyrosine phosphorylation in HUVECs. Also attenuates VEGFA-induced HUVEC cell migration and reduces A549 lung carcinoma cell viability in vitro. Inhibits binding of CendR motif in S1 protein of SARS-CoV-2 to NRP1, and reduces efficiency of SARS-CoV-2 infection in Caco-2 cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1210945-69-9
Quantity 10 mg
Solubility Soluble to 100 mM in DMSO and to 100 mM in 2eq. NaOH
Molecular Formula C17H19N7O5S3.CF3CO2H
Formula Weight 611.59
Product Type Antagonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.