missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EG-VEGF/PK1 Antibody (MM0043-2L13), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-93514R
This item is not returnable.
View return policy
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EG-VEGF/PK1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PROK1 | |
| human recombinant EG-VEGF protein | |
| 0.1 mL | |
| Primary | |
| This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0043-2L13 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
| Mouse | |
| Protein G purified | |
| Cell Cycle and Replication | |
| 84432 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction