missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EG-VEGF/PK1 Antibody (MM0043-2L13), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-93514JF646
This item is not returnable.
View return policy
Description
EG-VEGF/PK1 Monoclonal antibody specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EG-VEGF/PK1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
| human recombinant EG-VEGF protein | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 84432 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0043-2L13 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction