missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGF Antibody (008), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89582CL1
This item is not returnable.
View return policy
Description
EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for ELISA
Specifications
| EGF | |
| Monoclonal | |
| CoraFluor 1 | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EGF (Accession#: NP_001954.2; Asn 971-Arg 1023) . | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
| 1950 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 008 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction