missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGF Rabbit anti-Human, Clone: 8, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 541.00
Specifications
| Antigen | EGF |
|---|---|
| Clone | 8 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18225467
|
Novus Biologicals
NBP2-89582-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246566
|
Novus Biologicals
NBP2-89582-100UL |
100 μg |
€ 541.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGF Monoclonal specifically detects EGF in Human samples. It is validated for ELISA.Specifications
| EGF | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG | |
| Recombinant Human EGF protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 8 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| 1950 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title