Learn More
EGFL7 Mouse anti-Human, Clone: OTI2B5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807830
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Epidermal growth factor-like protein 7 (EGFL7), also known as VE-statin, MEGF7, Notch4-like protein or Zneu1, is a 29.6 kDa protein produced and secreted by endothelium. The protein is also expressed by some normal and maligt tissues. The main function of EGFL7 is to regulate angiogenesis and vasculogenesis. A high level of EFGL-7 can be found in tissues with active angiogenesis of a variety of disorders.Specifications
| EGFL7 | |
| Monoclonal | |
| Unconjugated | |
| EGFL7 | |
| NEU1, VE-STATIN, ZNEU1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51162 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2B5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UHF1 | |
| EGFL7 | |
| Human recombit protein fragment corresponding to amino acids 24-273 of human EGFL7 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.