missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eGFP Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06640APC
This item is not returnable.
View return policy
Description
eGFP Polyclonal antibody specifically detects eGFP in Nonspecies specific samples. It is validated for ELISA
Specifications
| eGFP | |
| Polyclonal | |
| ELISA | |
| Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein | |
| Produced in rabbits immunized with purified, recombinant eGFP (AFI57842.2; Met1-Lys239). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction