missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eGFP Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-59183
This item is not returnable.
View return policy
Description
eGFP Polyclonal antibody specifically detects eGFP in Nonspecies specific samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications
| eGFP | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1:800, Immunofluorescence 1:800 | |
| Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| Primary | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Non-species specific | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction