missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Antibody (DH8.3), Alexa Fluor™ 750 - Mutant, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50599AF750
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| EGFR | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EGFR | |
| Synthetic peptide corresponding to the junctional region of the truncated receptor LEEKKGNYVVTDHC,conjugated to keyhole limpet haemcyanin. | |
| Primary | |
| Clone DH8.3 of EGF R/ErbB1 antibody is specific for Truncated EGFR which is also called EGFR type III, EGFRvIII or delta-EGFR (characterized by a deletion of 267 amino acids in the extracellular domain, leading to a receptor which is unable to bind ligand yet is constitutively active). Because of its impaired internalisation and degradation, EGFRvIII is known to enhance tumourigenesis. NOTE: Clone DH8.3 DOES NOT cross-react with full length or wildtype EGFR (wtEGFR). | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| DH8.3 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Mouse | |
| 0.1 mL | |
| 1956 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction