missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Antibody (ICR10), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB600-724PCP
This item is not returnable.
View return policy
Description
EGFR Monoclonal antibody specifically detects EGFR in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot (Negative)
Specifications
| EGFR | |
| Monoclonal | |
| PerCP | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Extracellular domain of Human EGF-receptor from head and neck carcinoma. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
| 1956 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| ICR10 | |
| PBS | |
| Rat | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction