missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems EGFR-coupled magnetic beads
Description
The biotinylated EGFR protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-EGFR antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surface area and unique surface coating, which can help you get the best performance and highly reproducible results. This EGFR coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.
Specifications
Specifications
| Kit Contents | The conjugation was achieved by means of the binding between streptavidin and biotin. Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L Thus, the binding of streptavidin and bi |
| Content And Storage | -20°C, Avoid light |
| Shipping Condition | RT |
| Quantity | 10 mg |
| Type | Magnetic Affinity Bead |
| Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?