Learn More
Invitrogen™ EGFR (L858R Mutant) Recombinant Rabbit Monoclonal Antibody (RM380)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA544561
Product Code. 17846741
  • € 452.00 / 100µL
Description
EGFR (L858R Mutant) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
EGFR, epidermal growth factor receptor, is a receptor tyrosine kinases that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. EGFR is encoded by the EGFR gene located on chromosome 7 in humans. EGFR belongs to the HER/ERbB family of proteins that includes three other receptor tyrosine kinases, ERbB2, ERbB3, ERbB4. EGFR is a transmembrane receptor and binding of its cognate ligands such as EGF (Epidermal Growth Factor) and TGF alpha (Transforming Growth Factor alpha) to the extracellular domain leads to EGFR dimerization followed by autophosphorylation of the tyrosine residues in the cytoplasmic domain. Overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EGFR (L858R Mutant)
Recombinant Monoclonal
100 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
P00533
EGFR
A peptide corresponding to human EGFR (L858R Mutant Specific). The immunogen range is a synthetic peptide within to amino acids 800-900 of EGFR with L858R mutant.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
RM380
Unconjugated
EGFR
EGFR; Erbb1
Rabbit
Protein A
RUO
1956
-20°C