missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT2 Mouse anti-Human, Clone: OTI3E9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808820S
This item is not returnable.
View return policy
Description
SHMT2 Monoclonal antibody specifically detects SHMT2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SHMT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P34897 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6472 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E9 | |
| Unconjugated | |
| SHMT2 | |
| 2700043D08Rik; AA408223; AA986903; epididymis secretory sperm binding protein Li 51e; GLY A+; GLYA; glycine auxotroph A, human complement for hamster; glycine hydroxymethyltransferase; HEL-S-51e; mitochondrial serine hydroxymethyltransferase; serine aldolase; serine hydroxymethylase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 2; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, mitochondrial; Serine methylase; SHMT; SHMT 2; SHMT2; threonine aldolase | |
| Full length human recombinant protein of human SHMT2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction