missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR, p Tyr1173 Antibody (EM-13), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-44893AF532
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| EGFR | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EGFR | |
| Synthetic phospho-peptide covering sequence around tyrosine 1173 of human EGFR. | |
| Primary | |
| The antibody EM-13 reacts with human EGFR (ErbB1/ HER1) phosphorylated on tyrosine 1173. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| EM-13 | |
| Western Blot, Flow Cytometry | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Mouse | |
| 0.1 mL | |
| 1956 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction