missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGLN1/EGLN2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17197603
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197603 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17197603 Supplier Invitrogen™ Supplier No. PA5109677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DIRGDKITWI EGKEPGCETI GLLMSSMDDL IRHCNGKLGS YKINGRTKAM VACYPGNGTG YVRHVDNPNG DGRCVTCIYY LNKDWDAKVS GGILRIFPEG K.

Cellular oxygen sensor that catalyzes, under normoxic conditions, the post-translational formation of 4-hydroxyproline in hypoxia-inducible factor (HIF) alpha proteins. Hydroxylates a specific proline found in each of the oxygen-dependent degradation (ODD) domains (N-terminal, NODD, and C-terminal, CODD) of HIF1A. Also hydroxylates HIF2A. Has a preference for the CODD site for both HIF1A and HIF1B. Hydroxylated HIFs are then targeted for proteasomal degradation via the von Hippel-Lindau ubiquitination complex. Under hypoxic conditions, the hydroxylation reaction is attenuated allowing HIFs to escape degradation resulting in their translocation to the nucleus, heterodimerization with HIF1B, and increased expression of hypoxy-inducible genes. EGLN1 is the most important isozyme under normoxia and, through regulating the stability of HIF1, involved in various hypoxia-influenced processes such as angiogenesis in retinal and cardiac functionality. Target proteins are preferentially recognized via a LXXLAP motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGLN1/EGLN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Egln1
Gene Accession No. Q96KS0, Q9GZT9
Gene Alias 0610011A13Rik; AI503754; C1orf12; C85656; ECYT3; egl nine homolog 1; Egl nine homolog 2; egl nine-like protein 1; egl-9 family hypoxia inducible factor 1; egl-9 family hypoxia inducible factor 2; egl-9 family hypoxia-inducible factor 1; egl-9 family hypoxia-inducible factor 2; EGLN1; EGLN2; EIT6; EIT-6; Estrogen-induced tag 6; falkor; HALAH; HIF prolyl hydroxylase 2; HIF-1alpha prolyl-4-hydroxylase-1; Hif-p4h-1; Hif-p4h-2; HIFPH1; HIF-PH1; HIFPH2; HIF-PH2; HIF-prolyl hydroxylase 1; HIF-prolyl hydroxylase 2; HPH-1; HPH2; HPH-2; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; hypoxia-inducible factor prolyl hydroxylase 2; Ier4; immediate early response 4; ORF13; PHD1; PHD-1; PHD2; PHD-2; PNAS-118; PNAS-137; Prolyl hydroxylase domain-containing protein 1; prolyl hydroxylase domain-containing protein 2; SM20; SM-20; zinc finger MYND domain-containing protein 6; ZMYND6
Gene Symbols Egln1, EGLN2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112398, 54583
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.