missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody (2445B), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-76810MFV450
This item is not returnable.
View return policy
Description
EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| EGLN1/PHD2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2445B | |
| Flow Cytometry, Flow (Intracellular) | |
| C1orf12, EC 1.14.11.29, EC:1.14.11.291, ECYT3, egl nine homolog 1, egl nine-like protein 1, EGLN1, HALAH, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PNAS-118, PNAS-137, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6 | |
| This EGLN1/PHD2 antibody was developed against a partial synthetic peptide from the N-terminal portion of the human EGLN1/PHD2 protein. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Chromatin Research, Hypoxia, Immunology, Lipid and Metabolism | |
| 54583 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction