missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody (2445D), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-78782AF532
This item is not returnable.
View return policy
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EGLN1/PHD2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EGLN1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN1/PHD2 (amino acids 2-426) [UniProt #Q9QZT9] | |
| 0.1 mL | |
| Cancer, Chromatin Research, Hypoxia | |
| Human, Mouse |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2445D | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction