missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN2 Mouse anti-Human, Clone: OTI7H3, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808584S
This item is not returnable.
View return policy
Description
EGLN2 Monoclonal antibody specifically detects EGLN2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| EGLN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96KS0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 112398 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7H3 | |
| Unconjugated | |
| EGLN2 | |
| 0610011A13Rik; C85656; egl nine homolog 2; egl-9 family hypoxia inducible factor 2; egl-9 family hypoxia-inducible factor 2; Egln2; EIT6; EIT-6; Estrogen-induced tag 6; falkor; HIF-1alpha prolyl-4-hydroxylase-1; HIF-P4H 1; Hif-p4h-1; HIFPH1; HIF-PH1; HIF-prolyl hydroxylase 1; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; Ier4; immediate early response 4; P4H1; Phd1; PHD-1; Prolyl hydroxylase domain-containing protein 1; SM-20 | |
| Full length human recombinant protein of human EGLN2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction