missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN2/PHD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05472SS
This item is not returnable.
View return policy
Description
EGLN2/PHD1 Polyclonal antibody specifically detects EGLN2/PHD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| EGLN2/PHD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/mL, Immunocytochemistry/ Immunofluorescence 2 - 5 μg/mL | |
| DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1 | |
| Partial synthetic peptide made to an N-terminal portion of the human EGLN2/ PHD1 protein (between amino acids 10-75) [UniProt Q96KS0] | |
| 0.025 mg | |
| Cancer, Chromatin Research, Hypoxia, Signal Transduction | |
| 112398 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction