missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN3/PHD3 Antibody (EG188e/d5), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30440AF532
This item is not returnable.
View return policy
Description
EGLN3/PHD3 Monoclonal antibody specifically detects EGLN3/PHD3 in Human, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EGLN3/PHD3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| EG188e/d5 | |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
| Partial recombinant human PHD3/HIF Prolyl Hydroxylase 3 (UniProt# Q9H6Z9) | |
| 0.2 mL | |
| Apoptosis, Cancer, Chromatin Research, Hypoxia | |
| 112399 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction