Learn More
Lonza™ EGM™-Plus Endothelial Cell Growth Media-Plus BulletKit™
Description
- Culture kit containing growth medium and supplements.
- Developed to support optimal growth of human umbilical vein endothelial cells.
- Includes the following:
- 1 x EBM™ Plus Basal Medium (CC-5036), 475 mL
- 1 x EGM™Plus SingleQuots™ Supplement Pack (CC-4542) containing the following:
- 1 x Bottle FBS, 10.00 mL
- 1 x Vial Hydrocortisone, 0.50 mL
- 1 x Bottle L-Glutamine, 25.00 mL
- 1 x Vial EnGS, 1.00 mL
- 1 x Vial Ascorbic Acid, 0.50 mL
- 1 x Vial rhEGF, 0.50 mL
- 1 x Vial GA-1000, 0.50 mL
- 1 x Vial Heparin, 0.50 mL
Specifications
Specifications
| Cell Type | Endothelial Cells |
| Product Type | Growth Medium and Supplements |
| Quantity | Medium: 475 mL, FBS: 10.0 mL, Hydrocortisone: 0.50 mL, L-Glutamine: 25.0 mL, EnGS: 1.0 mL, Ascorbic Acid: 0.5 mL, rhEGF: 0.5 mL, GA-1000: 0.5 mL, Heparin: 0.50 mL |
| Content And Storage | Basalmedium, Fetales Kälberserum, Hydrocortison, L-Glutamin, EnGS, Ascorbinsäure, rhEGF, GA-1000, Heparin |