missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24657-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts in early endocytic membrane fusion and membrane trafficking of recycling endosomes. Plays a role in myoblast fusion. Recruited to endosomal membranes upon nerve growth factor stimulation, indirectly regulates neurite outgrowth.Specifications
| EHD1 | |
| Polyclonal | |
| Unconjugated | |
| EHD1 | |
| EH domain containing 1, EH domain containing protein 1, EHD1, H PAST, HPAST1, PAST, PAST homolog 1, PAST1, Testilin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10938, 13660, 293692 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q641Z6, Q9H4M9, Q9WVK4 | |
| EHD1 | |
| EHD1 Fusion Protein Ag18400 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction