missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EHD1 Recombinant Rabbit Monoclonal Antibody (5S4P9)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA542814

Product Code. 17719393

  • € 524.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive test controls include: HeLa, THP-1, U-251MG, Mouse liver, Mouse brain, Mouse spleen, Rat lung, Rat testis, Rat kidney. The target is usually found in the following locations: Cell membrane, Cell projection, Cytoplasmic side, Early endosome membrane, Peripheral membrane protein, Recycling endosome membrane, cilium membrane. Immunogen sequence: MFSWVSKDAR RKKEPELFQT VAEGLRQLYA QKLLPLEEHY RFHEFHSPAL EDADFDNKPM VLLVGQYSTG KTTFIRHLIE QDFPGMRIGP EPTTDSFIAV.

EHD1 acts in early endocytic membrane fusion and membrane trafficking of recycling endosomes. EHD1 also plays a role in myoblast fusion and is recruited to endosomal membranes upon nerve growth factor stimulation, indirectly regulates neurite outgrowth. Further, EHD1 belongs to a highly conserved gene family encoding EPS15 homology (EH) domain-containing proteins. The protein-binding EH domain was first noted in EPS15, a substrate for the epidermal growth factor receptor. The EH domain has been shown to be an important motif in proteins involved in protein-protein interactions and in intracellular sorting. The EHD1 protein is thought to play a role in the endocytosis of IGF1 receptors. Alternatively spliced transcript variants have been found for the EHD1 gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EHD1
Recombinant Monoclonal
1.28 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q641Z6, Q9H4M9, Q9WVK4
EHD1
Synthetic peptide
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
5S4P9
Unconjugated
EHD1
AA409636; CDABP0131; EH domain containing 1; EH domain-containing protein 1; Ehd1; EH-domain containing 1; H-PAST; HPAST1; mPAST1; PAST; PAST homolog 1; PAST1; RGD1306960; RME-1; testilin
Rabbit
Affinity chromatography
RUO
10938, 13660, 293692
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.