missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EHD4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16517331
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16517331 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16517331 Supplier Invitrogen™ Supplier No. PA589421

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MQEQLENYDF TKFHSLKPKL IEAVDNMLSN KISPLMNLIS QEETSTPTQL VQGGAFDGTT EGPFNQGYGE GAKEGADEEE WVVAKDKPVY DELFYTLSPI NGKISGVNAK KEMVTSKLPN SVLGKIWKLA DCDCDGMLDE EEFALAKHLI KIKLDGYELP SSLPPHLVPP SHRKSLPKAD; Positive Samples: HeLa, 293T, Mouse brain, Mouse heart; Cellular Location: Cell membrane, Cytoplasmic side, Early endosome membrane, Peripheral membrane protein, Recycling endosome membrane.

ATP- and membrane-binding protein that probably controls membrane reorganization/tubulation upon ATP hydrolysis. Plays a role in early endosomal transport. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHD4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EHD4
Gene Accession No. Q9EQP2, Q9H223
Gene Alias 2210022F10Rik; AI197390; AI846352; AV006278; EH domain containing 4; EH domain containing protein MPAST2; EH domain-containing protein 4; EHD4; EH-domain containing 4; FKSG7; HCA10; HCA11; Hepatocellular carcinoma-associated protein 10/11; hepatocellular carcinoma-associated protein HCA11; mPAST2; ortholog of rat pincher; PAST homolog 2; PAST homolog 4; Past2; PAST4; pincher; pinocytic chaperone
Gene Symbols EHD4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 362-541 of human EHD4 (NP_6446701).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30844, 98878
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.