missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EHMT2 Polyclonal Antibody
GREENER_CHOICE

Product Code. p-7184306
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17189413 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17189413 Supplier Invitrogen™ Supplier No. PA5111500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PRSEETLPKA TPDSLEPAGP SSPASVTVTV GDEGADTPVG ATPLIGDESE NLEGDGDLRG GRILLGHATK SFPSS Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 87%.

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. This gene is found near this cluster; it was mapped near the gene for C2 within a 120-kb region that included a HSP70 gene pair. These genes are all within the human major histocompatibility complex class III region. This gene was thought to be two different genes, NG36 and G9a, adjacent to each other but a recent publication shows that there is only a single gene. The protein encoded by this gene is thought to be involved in intracellular protein-protein interaction. There are three alternatively spliced transcript variants of this gene but only two are fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHMT2
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ehmt2
Gene Accession No. Q96KQ7
Gene Alias ankyrin repeat-containing protein; Bat8; C6orf30; D17Ertd710e; DKFZp686H08213; Ehmt2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; lysine N-methyltransferase 1C; NG36; NG36/G9a; Protein G9a
Gene Symbols Ehmt2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human EHMT2. Recombinant protein control fragment (Product #RP-105151).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10919
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.